Dividers for Slide Boxes
Leverancier: GLW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ZK25 ZK20 ZK15
GLWAZK25EA 2.5 EUR
GLWAZK25 GLWAZK20 GLWAZK15
Dividers for Slide Boxes
Objectglaasjeshouders
These slide box dividers are designed to store different slides and coverslips of various dimensions.
  • Adjustable length, pluggable into slide boxes
  • Can be used in in freezer down to −90 °C
For use with standard and special sized slides.
Opgelet: Not for use in autoclaves.