Microcentrifuge tube racks, SP Bel-Art
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F189080005 F189080015
BELAF189080005EA 93.2 EUR
BELAF189080005 BELAF189080015
Microcentrifuge tube racks, SP Bel-Art
Rekken Tube Racks
These microcentrifuge tube racks are made of lightweight PC and are stackable when loaded or empty. The locking clips firmly connect multiple racks together to form larger arrays that can be easily transported.
  • Durable and UV resistant
  • Maximise storage and save benchtop space
  • Securely holds 24 microcentrifuge tubes, either 0,5 ml or 1,5 to 2,0 ml
  • Can be combined when multiple tube sizes are used
  • Can be stored at temperatures down to –80 ºC
  • Steam autoclavable at 121 ºC