Magnetic stirrer heads with conical joint, for vaccum
Leverancier: JUCHHEIM Laborgerate GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10220.00 10230.00 10240.00
JUCH10220.00EA 915 EUR
JUCH10220.00 JUCH10230.00 JUCH10240.00
Magnetic stirrer heads with conical joint, for vaccum
Roerkoppen
Conical joint stirrer heads which have a stainless steel bottom, special steel shaft and chromium-plated brass upper body for vacuum.