Quartz, coarse wool 9-30 µm
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Silicium dioxide, Silicium (IV) oxide, Silicon dioxide, Sand, white quartz
393610100 393611000
ACRO393610100EA 33 EUR
ACRO393610100 ACRO393611000
Quartz, coarse wool 9-30 µm
Quartz
Formule: SiO₂
MW: 60.08 g/mol
Kookpunt: 2230
Smeltpunt: 1427
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00217788
CAS nummer: 14808-60-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5273273c3b2e473c6ced4e97e18a338f3d21302c&vid=9c414ebb18d830bfa8a77b318dbbb0e6a96da07b&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance White Coarse fibers