Sulphapyridine 95%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
N'-2-Pyridylsulphanilide, 4-Amino-N-(pyridin-2-yl)benzenesulphonamide, 2-(4-Aminobenzenesulphonamido)pyridine, 2-sulphanilamidopyridine
442850250
442851000
ACRO442850250EA
47.6
EUR
ACRO442850250
ACRO442851000
Sulphapyridine 95%
Sulphapyridine
Formule:
C₁₁H₁₁N₃O₂S MW: 249.29 g/mol Smeltpunt: 190 °C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00038036 CAS nummer: 144-83-2 |
Specification Test Results
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 190 to 193 °C |
Loss on drying | ≤0.5 % |
Sulfated ash | ≤0.1 % |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...