Evodiamine 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
457890010
457890050
ACRO457890010EA
61.4
EUR
ACRO457890010
ACRO457890050
Evodiamine 98%
Evodiamine
Formule:
C₁₉H₁₇N₃O Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD06407824 CAS nummer: 518-17-2 UN: 2811 ADR: 6.1,II |
Specification Test Results
Appearance | White to cream Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...