4-Chloro-1-butene 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 4-Chlorobut-1-ene
460160010 460160050
ACRO460160010EA 46.8 EUR
ACRO460160010 ACRO460160050
4-Chloro-1-butene 98%
4-Chloro-1-butene
Formule: C₄H₇Cl
MW: 90.55 g/mol
Kookpunt: 75
Dichtheid: 0.921 g/cm³
Vlampunt: –8 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00039386
CAS nummer: 927-73-1
UN: 1993
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4240 (20°C, 589 nm)
GC ≥97.5 %