Diethyl iminodicarboxylate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

460770010 460770050
ACRO460770010EA 32.2 EUR
ACRO460770010 ACRO460770050
Diethyl iminodicarboxylate 98%
Diethyl iminodicarboxylate
Formule: C₆H₁₁NO₄
Kookpunt: 132…134 °C (15 mmHg)
Smeltpunt: 51…53 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00031532
CAS nummer: 19617-44-8

Order Now

Specification Test Results

Appearance White to off-white Low melting solid
Infrared spectrum Conforms
Melting point 49 to 54 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR