Sealing ring
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRND6694EA 0 EUR
BRND6694
Sealing ring
Afdichtingen Afdichtringen
This sealing ring is made from FKM and is only suitable for autoclaving applications.