Vydac 302IC, anion exclusion HPLC columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
302IC4.6EA 1550 EUR
302IC4.6 302GD4/N
Vydac 302IC, anion exclusion HPLC columns
Kolommen voor chromatografie
Vydac 302IC is a low capacity anion-exchange (quaternary amine) material based on a high purity 10 µm large pore size silica.
  • Well referenced for 'non-suppressed' ion chromatography applications
  • Used with standard HPLC systems for analysis of common ions
  • Widely used for environmental analysis