Chemical resistant overalls, Body Plus T Best
Leverancier: BEST FRANCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01074T020M/L 01074T020XL 01074T020XXL
BESF01074T020M/LEA 0 EUR
BESF01074T020M/L BESF01074T020XL BESF01074T020XXL
Chemical resistant overalls, Body Plus T Best
Wegwerpkleding
These hooded overalls are made from non woven, spunbonded PP (SMS fabric).
  • Elasticated cuffs and back
  • Integrated boot covers
  • Zip closure with adhesive flap
  • Good moisture transfer
  • Permeable to air and water vapor, breathable
  • Filter 100% of particles larger than 3 microns
Weight: 50 g/m²