Sample bottles, glass
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5314-0100 5314-0010 5314-0025
BURK5314-0100EA 17.6 EUR
BURK5314-0100 300-0054 300-0055
Sample bottles, glass
Flessen Glazen flessen
Borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes