SP Bel-Art Freezer Storage Boxes, 81 Places
Leverancier: Bel-Art Products, a Part of SP
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F188520011 F188520016
BELAF188520011EA 146 EUR
BELAF188520011 BELAF188520016
SP Bel-Art Freezer Storage Boxes, 81 Places
Dozen Dozen voor cryobuisjes
These polypropylene storage boxes provide compact storage and keep cryo vials organized on the bench, in the freezer, or during transport.
  • Can be used at temperature ranges from –80 to +121 °C
  • Alphanumeric grid for tracking individual samples
  • 'No slide' features moulded in bottom and lid ensures secure stacking
  • Secure fitting, translucent lid allows to view the contents
  • Matte panels on two sides provides trouble-free labelling
Unlike cardboard models these sturdy boxes are autoclavable and will not break down with exposure to moisture and heavy use.
Opgelet: Do not fit in standard freezer racks.