Knitted caps, B013
Leverancier: Portwest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B013NAR B013BKR B013ORR B013YER B013GRR
PRTWB013NAREA 6.6 EUR
PRTWB013NAR PRTWB013BKR PRTWB013ORR PRTWB013YER PRTWB013GRR
Knitted caps, B013
Werkkleding
These caps are made from 100% acrylic knit with a 40 g Insulatex™ lining. Ideal for really cold days.
  • Insulatex™ lining ensures that the wearer remains warm
  • Available in a choice of colours