pH indicator, phenol red tablets and solutions, Lovibond®
Leverancier: Lovibond
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
471040 511750BT 511751BT 511770BT 511790BT 511791BT
634-8739EA 13.4 EUR
634-8739 LOVI511750BT LOVI511751BT LOVI511770BT LOVI511790BT LOVI511791BT
pH indicator, phenol red tablets and solutions, Lovibond®
Milieutestkits
Indicator for pH range 6,5 to 8,4.
Can be used for measuring pH in conjunction with the Comparator 2000+ system.