pH indicators, cresol red and cresol purple tablets, Lovibond®
Leverancier: Lovibond
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
511710BT 511711BT
LOVI511710BTEA 35.9 EUR
LOVI511710BT LOVI511711BT
pH indicators, cresol red and cresol purple tablets, Lovibond®
Milieutestkits
Indicator for pH range 7,2 to 8,8 and 1 to 2,6.
Can be used for measuring pH in conjunction with the Comparator 2000+ system.
Order Now
Specificaties
Environmentally Preferable