Frits, titanium, Unidense type
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-2FR8TI-5 JR-10FR8TI-5 JR-2FR6TI-5 JR-2FR4TI-5 JR-2FR1TI-5 JR-2FR1KTI-5 JR-2FR2TI-5 JR-.5FR1TI-5
JOURJR-2FR8TI-5EA 0 EUR
JOURJR-2FR8TI-5 JOURJR-10FR8TI-5 JOURJR-2FR6TI-5 JOURJR-2FR4TI-5 JOURJR-2FR1TI-5 JOURJR-2FR1KTI-5 JOURJR-2FR2TI-5 JOURJR-.5FR1TI-5
Frits, titanium, Unidense type
Filters Voorkolom filters en frits
Titanium frits are preferred over stainless steel frits for the application related to the analysis of sensitive substances such as biomolecules.
  • Biocompatible
  • Recommended for protein analysis
  • Higher corrosion resistance
  • Suitable for the solvents containing halides