Cleanroom mops, PocketMop™
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PKM6-14IR PKM11-14 PKM6-14 PKM6-18
MICRPKM6-14IREA 683 EUR
MICRPKM6-14IR MICRPKM11-14 MICRPKM6-14 MICRPKM6-18
Cleanroom mops, PocketMop™
Moppen
These mop can be used for controlled environments and is economical enough to compete with commercial mops that are often used in support or non-classified areas.
  • Lightweight and easy to maneuver
  • 100% smooth knit polyester
  • Compatible with strong disinfectants
The swivel joint rotates 360° but can be locked in a 180° movement for more precise, controlled cleaning. Economical enough for single use but can be laundered and reused.