Bottles, narrow neck, coated, with screw cap
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
612414345966 61.63241434595 61.63241432110 61.63241432125 61.63241432150 61.63241432194 612414321966 612414321968 612414345968 61632414345100 61.63241434525 61.63241434550 61.63241434594 612414345950
KAVA612414345966EA 1220 EUR
KAVA612414345966 KAVA61.63241434595 KAVA61.63241432110 KAVA61.63241432125 KAVA61.63241432150 KAVA61.63241432194 KAVA612414321966 KAVA612414321968 KAVA612414345968 KAVA61632414345100 KAVA61.63241434525 KAVA61.63241434550 KAVA61.63241434594 KAVA612414345950
Bottles, narrow neck, coated, with screw cap
Flessen Glazen flessen
These bottles are made of borosilicate glass 3.3, plastic coated, with screw cap.
  • Clear plastic coated to protect against mechanical breakage
  • Reduces the hazard of flying glass and liquid splashes in the event of breakage
  • Coating withstands steam sterilisation up to 135 °C