Volumetric flasks, class A, glass, clear, NS plastic stopper
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
936850 936852 936853 936854 936845 936838 936842 936843 936851 936849 936848 936846 936841
612-7114EA 51.8 EUR
612-7114 612-7119 612-7120 612-7121 612-7109 612-7105 612-7107 612-7108 612-7118 612-7113 612-7112 612-7110 612-7106
Volumetric flasks, class A, glass, clear, NS plastic stopper
Kolven Maatkolven
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, In)
  • Mark and inscriptions in ETERNA amber stain
  • DE-M marking
*Wide neck
Certificaten: DIN EN ISO 1042
Bestelinformatie: Supplied with an individual certificate. Volumetric flasks also available on request with batch certificate, a USP individual certificate or a DAkkS calibration certificate.