Storage vials, round, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRC0005 BRC0010
215-4118EA 230 EUR
215-4118 215-4119
Storage vials, round, Azlon®
flesjes
These are highly transparent PC vials with coloured PP cap. Ideal for sampling, production and packaging of steam sterilised biological solutions.
  • Leakproof and shatterproof
  • Suitable for repeated sterilisation
  • Withstands temperatures to –80 °C
Leveringsinformatie: Supplied with non sterile plastic wrapped trays with PP caps.