Centrifuge tubes, KIMAX®, KIMBLE®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SP45243-100 SP45243-200
SCERSP45243-100EA 710 EUR
SCERSP45243-100 SCERSP45243-200
Centrifuge tubes, KIMAX®, KIMBLE®
Buisjes Centrifugebuisjes
KIMAX® tube are designed for field use in testing petroleum.
  • Calibrated to contain
  • Scale and legend are durable white ceramic enamel
  • Short cone oil centrifuge tubes 152,4 mm
Order Now
Specificaties
Environmentally Preferable