GL transition fittings, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F761-16 F761-08
BOHLF761-16EA 75.7 EUR
BOHLF761-16 BOHLF761-08
GL transition fittings, BOLA
Fittingen en connectoren
Fitting made of PTFE, white.
  • Temperature resistance from –200 to +250 °C
  • Universal chemical resistance
  • FDA conform
One side with thread GL 14 for connecting hard-walled tubing and tubes by means of BOLA laboratory screw joints. Other side with female thread UNF 1/4" 28G for connecting flanged tubing with O. D. 1,6 and 3,2 mm by means of tube end fittings.