Tri-camp GL-stopcocks, BOLA
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D917-42 D917-02
BOHLD917-42EA 233 EUR
BOHLD917-42 BOHLD917-02
Tri-camp GL-stopcocks, BOLA
Afsluitkranen
Made of PTFE/PP.
  • Very good chemical resistance
  • FDA confirmity
  • Temperature resistant from 0 to +110 °C
  • Quick and easy disconnection of flow
Cylindrical 2-Way stopcock with straight bore, Tri-Clamp and GL-threaded neck, made of PTFE for reliable tightness and grip made of PP for marking the flow direction.
Certificaten: DIN 32 676