Methylprednisolone
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 6α-Methylprednisolone
338800010 338800050
ACRO338800010EA 72.4 EUR
ACRO338800010 ACRO338800050
Methylprednisolone
Methylprednisolone
Formule: C₂₂H₃₀O₅
MW: 374.48 g/mol
Smeltpunt: 227…231 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00010591
CAS nummer: 83-43-2
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥96.0 %
Specific optical rotation +97° to +103° (20°C, 589 nm) (1 % in ethanol)