Micro particle size standard, polystyrene monodisperse, analytical, 0,1 µm, NIST, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
90517-10ML-F 90517-5ML-F
90517-10ML-FEA 425 EUR
90517-10ML-F 90517-5ML-F
Micro particle size standard, polystyrene monodisperse, analytical, 0,1 µm, NIST, Supelco®
Standaarden Organische standaarden
Particle size and standard deviation are determined with an accuracy of 0,001 μm.
  • Concentration: 2% (solids)
  • Format: neat
  • Form: aqueous suspension
Size determination of these particles follows procedures described by National Institute of Standards and Technology (NIST) respectively by the Community Bureau of Reference (BCR).
Certificaten: Traceable to NIST and BCR particle size standard.