Extractors for solids
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05431031 05431037 05431049
LENZ05431031EA 235 EUR
LENZ05431031 LENZ05431037 LENZ05431049
Extractors for solids
Extractors Soxhlet-extractors en Soxhlet-type extractors
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of solids (warm).
The complete apparatuses are consisting of:
Extractor head with glass thimbles for solids, Dimroth condenser, with thread GL 14, round bottom flask.