Plugs and caps
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-408 JR-410 JR-409 JR-ZC1PK JR-CCPK-1 JR-ZC.5FPK JR-ZP.5FPK
JOURJR-408EA 16.1 EUR
JOURJR-408 JOURJR-410 JOURJR-409 JOURJR-ZC1PK JOURJR-CCPK-1 JOURJR-ZC.5FPK JOURJR-ZP.5FPK
Plugs and caps
Fittingen en connectoren
These plugs and caps are used for plugging ports and capping solvent lines.
  • Good chemical resistance
  • Pressure rating: <350 bar