HYDRANAL™ humidity absorber for Karl Fischer titration
Leverancier: Honeywell Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34788-500G
34788-1KG
34788-6X1KG
34788-500GEA
57.3
EUR
34788-500G
34788-1KG
34788-6X1KG
HYDRANAL™ humidity absorber for Karl Fischer titration
Karl Fischer titranten en reagentia
HYDRANAL™ Humidity absorber is a drying agent for air and gases for Karl Fischer titration (with indicator).
- Water absorption capacity: 25%
- Regeneration: 140 °C (until the colour turns back to orange)