Probes for precision digital thermometer
Leverancier: Amarell
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E910650 E910654 E910656 E910652
AMARE910650EA 56.4 EUR
AMARE910650 AMARE910654 AMARE910656 AMARE910652
Probes for precision digital thermometer
Temperatuursondes
Precise Pt 100 class A probe of stainless steel, PVC-cable.