Restriction enzymes, PstI, FastDigest™, Fermentas
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FD0614 FD0615
FD0614EA 72.25 EUR
FD0614 FD0615
Restriction enzymes, PstI, FastDigest™, Fermentas
Reagentia voor nucleïnezuren Restriction Enzymes
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications
Designed to complete digestion in 5 to 15 minutes.