Restriction enzymes, HindIII, FastDigest™, Fermentas
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FD0504
FD0505
FD0504EA
53.25
EUR
FD0504
FD0505
Restriction enzymes, HindIII, FastDigest™, Fermentas
Reagentia voor nucleïnezuren
Restriction Enzymes
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
- 100% activity of all FastDigest™ enzymes in one universal reaction buffer
- Direct loading on gels with FastDigest™ Green buffer
- 100% buffer compatibility with downstream applications
Designed to complete digestion in 5 to 15 minutes.