Anti-PKC alpha Mouse Monoclonal Antibody
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ADIKAMPK020D ADIKAMPK020F
ENZOADIKAMPK020DEA 275 EUR
ENZOADIKAMPK020D ENZOADIKAMPK020F
Anti-PKC alpha Mouse Monoclonal Antibody
Antilichamen
Protein Kinase C (PKC) is a large superfamily of serine/threonine kinases that mediate essential cellular signals required for activation, proliferation, differentiation and survival. There are at least ten PKC isotypes that are closely related in structure but that have distinct patterns of tissue distribution and function. The PKC isotypes can be subdivided into three classes based on primary structure and biochemical properties. These are: classical or conventional PKC isotypes (cPKC), novel PKC isotypes (nPKC) and atypical PKC isotypes (aPKC).
  • For Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse, rat, bovine, dog, fish, gerbil, monkey, pig, rabbit and sheep PKCalpha. Detects a band of ~80kDa by Western blot.
Type: Primary
Antigen: PKC ALPHA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human