Anti-Ubiquitin Mouse Monoclonal Antibody

Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ADISPA203D ADISPA203F
ENZOADISPA203DEA 299 EUR
ENZOADISPA203D ENZOADISPA203F
Anti-Ubiquitin Mouse Monoclonal Antibody
Antilichamen
Ubiquitin (Ub) plays a very important role in regulated non-lysosomal ATP dependent protein degradation. The protein to be degraded is conjugated to Ub and the ubiquinated protein is then selectively degraded by the 26S complex, a multicatalytic cytosolic and nuclear protease. The Ub-proteasome proteolytic pathway, which is a complex process, is implicated to be of great importance for regulating numerous cellular processes.

  • For Western Blot

Recognizes human, mouse, rat, bovine, hamster, and rabbit Ubiquitin. Detects a band of ~6.5kDa by Western blot.

Type: Primary
Antigen: UBIQUITIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR