Anti-S Mouse Monoclonal Antibody

Leverancier: Enzo Life Sciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

ENZOALX804164C100EA 495 EUR
ENZOALX804164C100 ALEXALX804164C100
Anti-S Mouse Monoclonal Antibody
Antilichamen
Host: Mouse, Isotype: IgM

Recognizes a broad range of serine-phosphorylated proteins in crude cell extracts, interacting with phosphoserine surrounded by positive or neutral aa (e.g. substrates of PKA, PKB, PKC, PKG etc.).Please note that phosphoserine detection by MAbs always depends on the surrounding aa sequence.

For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot

Reconstitute with 1ml water (15 minutes at room temperature).

Bestelinformatie: Positive control included.

Type: Primary
Antigen: S
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR