Anti-S Mouse Monoclonal Antibody
Leverancier: Enzo Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALEXALX804167C100EA 799 EUR
ALEXALX804167C100 ENZOALX804167C100
Anti-S Mouse Monoclonal Antibody
Antilichamen
Host: Mouse, Isotype: IgM
Recognizes a broad range of serine-phosphorylated proteins in crude cell extracts, strongly interacting with pSK and pSP motifs, thus specifically recognizing substrates of MAP/SAP kinases and CDC kinases.Please note that phosphoserine detection by MAbs always depends on the surrounding aa sequence.
For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Reconstitute with 1ml water (15 minutes at room temperature).
Bestelinformatie: Positive control included.
Type: Primary
Antigen: S
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: pan