Anti-p47Phox, NCF1 Rabbit Polyclonal Antibody
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ADIOSA5500050 ADIOSA5500200
ENZOADIOSA5500050EA 245 EUR
ENZOADIOSA5500050 ENZOADIOSA5500200
Anti-p47Phox, NCF1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: p47Phox, NCF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse