Antifade mounting medium
Leverancier: ENZO LIFE SCIENCES
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ENZ53002M010
ENZ53003M010
ENZOENZ53002M010EA
165
EUR
ENZOENZ53002M010
ENZOENZ53003M010
Antifade mounting medium
Kleurstoffen
Microscopy and staining reagents
Antifade mounting medium (hardset) is an antifade reagent that causes little or no quenching. Use of the mounting medium significantly extends the photobleaching half-life of the fluorescent stain.
- Inhibits photobleaching of fluorescent dyes and fluorescent proteins
- Hardening formulation
Recommended for use on fixed cells and tissues that have been stained with fluorescent stains. Cells can be viewed immediately, or if long term storage is desired, the reagent can be allowed to harden overnight.