IL-10 (human) ELISA Kit
Leverancier: ENZO LIFE SCIENCES
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ADI900036
ADI901036
ENZOADI900036EA
684
EUR
ENZOADI900036
ENZOADI901036
IL-10 (human) ELISA Kit
Testen
ELISAs
The IL-10 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit kor the quantitative determination of human IL-10 in culture supernatants, plasma, and serum.
- Ready to use, liquid colour coded reagents and pre-coated plates reduce errors and save time
- Sensitive - detect <4 pg/ml of IL-10
- Results in <3 hours from up to 39 samples in duplicate
Leveringsinformatie: Kit includes microtiter plate, conjugate, antibody, assay buffer 13 concentrate, wash buffer concentrate, standard, TMB substrate, stop solution 2.