GFP-Certified™ FluoForte™ Reagent
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ENZ520165C50 ENZ52016M001
ENZOENZ520165C50EA 376 EUR
ENZOENZ520165C50 ENZOENZ52016M001
GFP-Certified™ FluoForte™ Reagent
Koppelings- en labelingssystemen voor eiwitten
Calcium dye
The GFP-Certified™ FluoForte™ Reagent is the brightest and most sensitive fluorescent calcium indictor relative to other commercially available red fluorescent dyes with maximum excitation at ~530 nm and maximum emission at ~555 nm. This product is used in a homogeneous fluorescence-based assay for detecting intracellular calcium mobilization across a broad spectrum of biological targets.