beta-Amyloid (1-42)
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX151002P001 ALX151002PC05
ENZOALX151002P001EA 1140 EUR
ENZOALX151002P001 ENZOALX151002PC05
beta-Amyloid (1-42)
Eiwitten en peptiden
For increased cell permeability the peptide was covalently linked to the 10 aa recognized by the TAT transporter.
Bestelinformatie: Quantity: *Sold as net peptide weight.