beta-Amyloid (1-40)
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX151003P001 ALX151003PC05
ENZOALX151003P001EA 1060 EUR
ENZOALX151003P001 ENZOALX151003PC05
beta-Amyloid (1-40)
Eiwitten en peptiden
Bestelinformatie: Quantity: *Sold as net peptide weight.
Order Now
Specificaties
Protein/peptide name beta-Amyloid (1-40) beta-Amyloid (1-40)
Environmentally Preferable
Species
Bron
Zuiverheid >97%