N-Acetyl-Asp-Glu-OH
Leverancier: ENZO LIFE SCIENCES
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX151020M010
ALX151020M050
ENZOALX151020M010EA
66
EUR
ENZOALX151020M010
ENZOALX151020M050
N-Acetyl-Asp-Glu-OH
Eiwitten en peptiden
Glutamate receptor ligand
Endogenous neurotransmitter with high affinity for the brain glutamate receptor. N-acetylated α-linked acidic dipeptidase (NAALADase), a membrane-bound peptidase, hydrolyzes α-NAAG, a major brain peptide, to N-acetylaspartate and glutamate. Discriminates between mGluR2 and mGluR3. mGluR3 activator. NMDA receptor antagonist. Neuroprotective.