Human recombinant Annexin V, FITC (Fluorescein Isothiocyanate)
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX2092563100 ALX209256T100
ENZOALX2092563100EA 681 EUR
ENZOALX2092563100 ENZOALX209256T100
Human recombinant Annexin V, FITC (Fluorescein Isothiocyanate)
Eiwitten en peptiden
FITC-labelled recombinant human annexin V fusion protein (produced in E. coli) shows bright green fluorescence (Ex(max): 488 nm; Em(max): 530 nm).
Bestelinformatie: Quantity: Sufficient for 100 or 300 tests. 1 test is equivalent to 1μg of the conjugated protein in an experimental volume of 0.4 ml.
Order Now
Specificaties
Protein/peptide name Annexin V Annexin V
Environmentally Preferable
Protein/peptide type Recombinant Recombinant
Species Human Human
Bron
Conjugatie FITC (Fluoresceïne-isothiocyanaat) FITC (Fluoresceïne-isothiocyanaat)
Zuiverheid >90%
Concentratie 1 mg/ml
Formulering Liquid. In PBS, pH 7,4, containing 0,02% sodium azide.
Purification Affinity purified