Ac-IETD-pNA
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX260045M001 ALX260045M005
ENZOALX260045M001EA 84.7 EUR
ENZOALX260045M001 ENZOALX260045M005
Ac-IETD-pNA
Eiwitten en peptiden
Caspase and granzyme substrate
Chromogenic substrate for caspase-6, -8 and granzyme B. Also a substrate for caspase-10. Similar to Ac-IETD-AMC and Ac-IETD-AFC but cleavage is monitored colorimetrically by absorbance at 405 nm.