Ac-LEVD-AFC
Leverancier: ENZO LIFE SCIENCES
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX260084M001
ALX260084M005
ENZOALX260084M001EA
93.5
EUR
ENZOALX260084M001
ENZOALX260084M005
Ac-LEVD-AFC
Eiwitten en peptiden
Caspase-4 substrate
Fluorogenic substrate for caspase-4. Similar to the fluorogenic substrate Ac-LEVD-AMC (Prod. No. ALX-260-083); the AFC fluorophore is more sensitive.