Ac-LETD-AFC
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX260118M005 ALX260118M010
ENZOALX260118M005EA 316 EUR
ENZOALX260118M005 ENZOALX260118M010
Ac-LETD-AFC
Eiwitten en peptiden
Caspase-8 substrate
Fluorogenic substrate for caspase-8.