OmniCathepsin™ fluorogenic Substrate
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BMLP1390010 BMLP1390050
ENZOBMLP1390010EA 165 EUR
ENZOBMLP1390010 ENZOBMLP1390050
OmniCathepsin™ fluorogenic Substrate
Eiwitten en peptiden
Fluorogenic substrate cleaved by plasma kallikrein and kallikrein 8, but not plasmin; by papain; and by most cathepsins (cathepsins B, C/DPP-I, F, K/O2, L, L2/V, O, S, and X/Z). Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450.
  • Substrate is useful for inhibitor screening and kinetic analysis
  • Also available: fluorogenic calibration standard AMC (BML-KI144)