Salmon Calcitonin
Leverancier: ENZO LIFE SCIENCES
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BMLP4200001
BMLP4200005
ENZOBMLP4200001EA
139
EUR
ENZOBMLP4200001
ENZOBMLP4200005
Salmon Calcitonin
Eiwitten en peptiden
Reduces blood calcium
Calcium-regulating peptide hormone released from the thyroid. Lowers calcium concentration in the blood and decreases bone resorption by osteoclasts. Used for therapeutic applications in osteoporosis. Salmon calcitonin peptide sequence differs from mammalian but is more potent.