Z-VAD-FMK
Leverancier: ENZO LIFE SCIENCES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ALX260020M001 ALX260020M005
ENZOALX260020M001EA 209 EUR
ENZOALX260020M001 ENZOALX260020M005
Z-VAD-FMK
Eiwitten en peptiden
Caspase inhibitor
Irreversible, cell permeable broad-spectrum caspase inhibitor.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21782967a806888623cffa35edfc1bcb22e391a0&vid=6be0cd0c7ce932916047ee79ce47c51c959ad88d&prid=web&clid=SAPDAM
Order Now
Specificaties
Protein/peptide name Z-VAD-FMK
Environmentally Preferable
Species
Bron
Zuiverheid >92% (HPLC)